Assay Details
| Gene Symbol: NR2E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003353279.4 | XP_003353327.1 | 5799 | 123 | ||
| GenBank mRNA | AK391420.1 | - | 1 - 2 | 50 | 123 | |
More Information
Set Membership: |
|

NR2E1
Genomic Map