Assay Details
| Gene Symbol: SMDT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003481528.3 | XP_003481576.1 | 367 | 87 | ||
| GenBank mRNA | AK391680.1 | - | 1 - 2 | 225 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SMDT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003481528.3 | XP_003481576.1 | 367 | 87 | ||
| GenBank mRNA | AK391680.1 | - | 1 - 2 | 225 | 87 | |
Set Membership: |
|