Assay Details
| Gene Symbol: SMDT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003481528.3 | XP_003481576.1 | 508 | 108 | ||
| GenBank mRNA | AK391680.1 | - | 2 - 3 | 366 | 108 | |
More Information
Set Membership: |
|

SMDT1
Genomic Map