Assay Details
| Gene Symbol: VAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005653160.3 | XP_005653217.1 | 476 | 102 | ||
| GenBank mRNA | AK391781.1 | - | 2 - 3 | 273 | 102 | |
More Information
Set Membership: |
|

VAMP1
Genomic Map