Assay Details
| Gene Symbol: VSNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021087821.1 | XP_020943480.1 | 367 | 55 | ||
| GenBank mRNA | AK391844.1 | - | 1 - 2 | 252 | 55 | |
| AK399876.1 | - | 1 - 2 | 267 | 55 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021087821.1 transcripts.
Set Membership: |
|

VSNL1
Genomic Map