Assay Details
| Gene Symbol: VSNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021087821.1 | XP_020943480.1 | 532 | 178 | ||
| GenBank mRNA | AK391844.1 | - | 2 - 3 | 417 | 178 | |
| AK399876.1 | - | 2 - 3 | 432 | 178 | ||
More Information
Set Membership: |
|

VSNL1
Genomic Map