Assay Details
| Gene Symbol: UBXN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001924982.7 | XP_001925017.1 | 881 | 78 | ||
| GenBank mRNA | AK391877.1 | - | 7 - 8 | 848 | 78 | |
More Information
Set Membership: |
|

UBXN2B
Genomic Map