Assay Details
| Gene Symbol: TMEM158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358398.4 | XP_003358446.1 | 617 | 97 | ||
| GenBank mRNA | AK392187.1 | - | 1 | 420 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM158| Gene Symbol: TMEM158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358398.4 | XP_003358446.1 | 617 | 97 | ||
| GenBank mRNA | AK392187.1 | - | 1 | 420 | 97 | |
Set Membership: |
|