Assay Details
Gene Symbol: IFI6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021095658.1 | XP_020951317.1 | 329 | 77 | ||
XM_021095659.1 | XP_020951318.1 | 319 | 77 | |||
XM_021095660.1 | XP_020951319.1 | 331 | 77 | |||
GenBank mRNA | AK392512.1 | - | 3 - 4 | 254 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
