Assay Details
| Gene Symbol: RNF128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003135291.6 | XP_003135339.3 | 3053 | 102 | ||
| XM_021079706.1 | XP_020935365.1 | 2587 | 102 | |||
| GenBank mRNA | AK392653.1 | - | 1 | 1325 | 102 | |
More Information
Set Membership: |
|

RNF128
Genomic Map