Assay Details
| Gene Symbol: SLX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013995704.2 | XP_013851158.2 | 2086 | 64 | ||
| XM_021088081.1 | XP_020943740.1 | 2119 | 64 | |||
| XM_021088082.1 | XP_020943741.1 | 1862 | 64 | |||
| XM_021088083.1 | XP_020943742.1 | 1898 | 64 | |||
| GenBank mRNA | AK392697.1 | - | 9 - 10 | 1874 | 64 | |
More Information
Set Membership: |
|

SLX4
Genomic Map