Assay Details
| Gene Symbol: ATP6V1C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125387.4 | XP_003125435.1 | 320 | 80 | ||
| XM_005655276.3 | XP_005655333.1 | 320 | 80 | |||
| XM_021087842.1 | XP_020943501.1 | 321 | 80 | |||
| XM_021087843.1 | XP_020943502.1 | 383 | 80 | |||
| XM_021087844.1 | XP_020943503.1 | 506 | 80 | |||
| XM_021087845.1 | XP_020943504.1 | 316 | 80 | |||
| XM_021087846.1 | XP_020943505.1 | 247 | 80 | |||
| XM_021087847.1 | XP_020943506.1 | 320 | 80 | |||
| GenBank mRNA | AK393316.1 | - | 3 - 4 | 301 | 80 | |
More Information
Set Membership: |
|

ATP6V1C2
Genomic Map