Assay Details
| Gene Symbol: ARID1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095630.1 | XP_020951289.1 | 5442 | 75 | ||
| XM_021095631.1 | XP_020951290.1 | 5438 | 75 | |||
| GenBank mRNA | AK393992.1 | - | 1 | 220 | 75 | |
More Information
Set Membership: |
|

ARID1A
Genomic Map