Assay Details
| Gene Symbol: CHST9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096199.1 | XP_020951858.1 | 2952 | 89 | ||
| XM_021096200.1 | XP_020951859.1 | 3229 | 89 | |||
| XM_021096201.1 | XP_020951860.1 | 3176 | 89 | |||
| XM_021096202.1 | XP_020951861.1 | 2487 | 89 | |||
| GenBank mRNA | AK395701.1 | - | 5 | 2916 | 89 | |
More Information
Set Membership: |
|

CHST9
Genomic Map