Assay Details
| Gene Symbol: DNAJC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001925120.7 | XP_001925155.3 | 247 | 102 | ||
| GenBank mRNA | AK395991.1 | - | 1 - 2 | 150 | 102 | |
| AY609568.1 | - | 1 - 2 | 118 | 102 | ||
More Information
Set Membership: |
|

DNAJC15
Genomic Map