Assay Details
| Gene Symbol: NUAK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126093.6 | XP_003126141.3 | 5036 | 85 | ||
| XM_013988236.2 | XP_013843690.1 | 4790 | 85 | |||
| XM_013988237.2 | XP_013843691.1 | 4835 | 85 | |||
| XM_021091582.1 | XP_020947241.1 | 7094 | 85 | |||
| XM_021091583.1 | XP_020947242.1 | 4919 | 85 | |||
| GenBank mRNA | AK396215.1 | - | 1 | 1019 | 85 | |
More Information
Set Membership: |
|

NUAK1
Genomic Map