Assay Details
| Gene Symbol: CREB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013985747.2 | XP_013841201.2 | 4151 | 106 | ||
| XM_013985749.2 | XP_013841203.2 | 708 | 106 | |||
| XM_021078565.1 | XP_020934224.1 | 3750 | 106 | |||
| XM_021078566.1 | XP_020934225.1 | 4151 | 106 | |||
| XM_021078567.1 | XP_020934226.1 | 1019 | 106 | |||
| XM_021078568.1 | XP_020934227.1 | 655 | 106 | |||
| XM_021078569.1 | XP_020934228.1 | 399 | 106 | |||
| GenBank mRNA | AK396364.1 | - | 6 - 7 | 670 | 106 | |
More Information
Set Membership: |
|

CREB5
Genomic Map