Assay Details
| Gene Symbol: SPPL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928829.7 | XP_001928864.5 | 1315 | 62 | ||
| XM_021072704.1 | XP_020928363.1 | 1013 | 62 | |||
| XM_021072705.1 | XP_020928364.1 | 1427 | 62 | |||
| GenBank mRNA | AK396732.1 | - | 8 - 9 | 1320 | 62 | |
More Information
Set Membership: |
|

SPPL3
Genomic Map