Assay Details
| Gene Symbol: CDCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005669452.3 | XP_005669509.1 | 926 | 67 | ||
| XM_013981558.2 | XP_013837012.1 | 760 | 67 | |||
| GenBank mRNA | AK396753.1 | - | 3 - 4 | 692 | 67 | |
More Information
Set Membership: |
|

CDCP1
Genomic Map