Assay Details
| Gene Symbol: EFNA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013997020.2 | XP_013852474.2 | 546 | 62 | ||
| GenBank mRNA | AK397006.1 | - | 1 - 2 | 187 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: EFNA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013997020.2 | XP_013852474.2 | 546 | 62 | ||
| GenBank mRNA | AK397006.1 | - | 1 - 2 | 187 | 62 | |
Set Membership: |
|