Assay Details
| Gene Symbol: LATS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005659149.3 | XP_005659206.2 | 5615 | 93 | ||
| GenBank mRNA | AK397274.1 | - | 1 | 1423 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: LATS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005659149.3 | XP_005659206.2 | 5615 | 93 | ||
| GenBank mRNA | AK397274.1 | - | 1 | 1423 | 93 | |
Set Membership: |
|