Assay Details
| Gene Symbol: NYNRIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005666233.3 | XP_005666290.3 | 7339 | 88 | ||
| GenBank mRNA | AK397395.1 | - | 1 | 1321 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: NYNRIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005666233.3 | XP_005666290.3 | 7339 | 88 | ||
| GenBank mRNA | AK397395.1 | - | 1 | 1321 | 88 | |
Set Membership: |
|