Assay Details
| Gene Symbol: SIRPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021077411.1 | XP_020933070.1 | 1014 | 83 | ||
| XM_021077412.1 | XP_020933071.1 | 1070 | 83 | |||
| GenBank mRNA | AK397720.1 | - | 4 - 5 | 718 | 83 | |
More Information
Set Membership: |
|

SIRPB2
Genomic Map