Assay Details
| Gene Symbol: SLC16A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013989834.2 | XP_013845288.2 | 1379 | 80 | ||
| XM_021066361.1 | XP_020922020.1 | 2130 | 80 | |||
| XM_021066362.1 | XP_020922021.1 | 1573 | 80 | |||
| XM_021066363.1 | XP_020922022.1 | 1570 | 80 | |||
| XM_021066364.1 | XP_020922023.1 | 2150 | 80 | |||
| XM_021066365.1 | XP_020922024.1 | 2122 | 80 | |||
| XM_021066366.1 | XP_020922025.1 | 1605 | 80 | |||
| XM_021066367.1 | XP_020922026.1 | 1434 | 80 | |||
| XM_021066368.1 | XP_020922027.1 | 1954 | 80 | |||
| GenBank mRNA | AK397753.1 | - | 1 - 2 | 299 | 80 | |
| EU650276.1 | - | 1 - 2 | 574 | 80 | ||
More Information
Set Membership: |
|

SLC16A3
Genomic Map