Assay Details
| Gene Symbol: CORO1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021082756.1 | XP_020938415.1 | 1081 | 79 | ||
| GenBank mRNA | AK399062.1 | - | 3 - 4 | 326 | 79 | |
| AK399145.1 | - | 7 - 8 | 926 | 79 | ||
More Information
Set Membership: |
|

CORO1B
Genomic Map