Assay Details
| Gene Symbol: ETAA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021087430.1 | XP_020943089.1 | 5500 | 90 | ||
| XM_021087433.1 | XP_020943092.1 | 5413 | 90 | |||
| XM_021087434.1 | XP_020943093.1 | 5060 | 90 | |||
| XM_021087435.1 | XP_020943094.1 | 8203 | 90 | |||
| GenBank mRNA | AK399581.1 | - | 1 | 1720 | 90 | |
More Information
Set Membership: |
|

ETAA1
Genomic Map