Assay Details
| Gene Symbol: PRTN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013994800.2 | XP_013850254.1 | 3506 | 89 | ||
| GenBank mRNA | AK399742.1 | - | 1 - 2 | 28 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PRTN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013994800.2 | XP_013850254.1 | 3506 | 89 | ||
| GenBank mRNA | AK399742.1 | - | 1 - 2 | 28 | 89 | |
Set Membership: |
|