Assay Details
| Gene Symbol: NDUFAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021077006.1 | XP_020932665.1 | 316 | 134 | ||
| GenBank mRNA | AY609396.1 | - | 2 - 3 | 248 | 134 | |
More Information
Set Membership: |
|

NDUFAF2
Genomic Map