Assay Details
| Gene Symbol: LAMP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926375.7 | XP_001926410.3 | 2330 | 64 | ||
| XM_021077839.1 | XP_020933498.1 | 1455 | 64 | |||
| GenBank mRNA | AY803096.1 | - | 1 | 323 | 64 | |
More Information
Set Membership: |
|

LAMP5
Genomic Map