Assay Details
| Gene Symbol: TDO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003128997.6 | XP_003129045.3 | 698 | 72 | ||
| GenBank mRNA | FJ810127.1 | - | 2 - 3 | 213 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: TDO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003128997.6 | XP_003129045.3 | 698 | 72 | ||
| GenBank mRNA | FJ810127.1 | - | 2 - 3 | 213 | 72 | |
Set Membership: |
|