Assay Details
| Gene Symbol: GNAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021063492.1 | XP_020919151.1 | 1255 | 81 | ||
| XM_021063493.1 | XP_020919152.1 | 1871 | 81 | |||
| GenBank mRNA | FR715044.1 | - | 2 - 3 | 285 | 81 | |
More Information
Set Membership: |
|

GNAT3
Genomic Map