Assay Details
| Gene Symbol: NLRP12L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005657910.3 | XP_005657967.1 | 3152 | 74 | ||
| XM_013990988.2 | XP_013846442.1 | 3073 | 74 | |||
| XM_013990989.2 | XP_013846443.1 | 3215 | 74 | |||
| XM_013990990.2 | XP_013846444.1 | 3053 | 74 | |||
| XM_013990991.2 | XP_013846445.1 | 3122 | 74 | |||
| XM_013990992.2 | XP_013846446.1 | 3172 | 74 | |||
| XM_013990993.2 | XP_013846447.1 | 3197 | 74 | |||
| XM_013990994.2 | XP_013846448.1 | 2893 | 74 | |||
| XM_021079742.1 | XP_020935401.1 | 3246 | 74 | |||
| XM_021079744.1 | XP_020935403.1 | 3206 | 74 | |||
| XM_021079745.1 | XP_020935404.1 | 3073 | 74 | |||
| XM_021079746.1 | XP_020935405.1 | 2927 | 74 | |||
| XM_021079747.1 | XP_020935406.1 | 3069 | 74 | |||
| XM_021079748.1 | XP_020935407.1 | 2856 | 74 | |||

NLRP12L
Genomic Map