Assay Details
| Gene Symbol: SLC36A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134140.5 | XP_003134188.2 | 521 | 76 | ||
| XM_021077072.1 | XP_020932731.1 | 592 | 76 | |||
| XM_021077073.1 | XP_020932732.1 | 327 | 76 | |||
More Information
Set Membership: |
|

SLC36A1
Genomic Map