Assay Details
| Gene Symbol: SLC17A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121410.4 | XP_003121458.2 | 1113 | 75 | ||
| XM_005654403.3 | XP_005654460.1 | 875 | 75 | |||
| XM_013985139.2 | XP_013840593.1 | 1109 | 75 | |||
More Information
Set Membership: |
|

SLC17A5
Genomic Map