Assay Details
| Gene Symbol: SNX20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094194.1 | XP_020949853.1 | 630 | 87 | ||
| XM_021094195.1 | XP_020949854.1 | 449 | 87 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SNX20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094194.1 | XP_020949853.1 | 630 | 87 | ||
| XM_021094195.1 | XP_020949854.1 | 449 | 87 | |||
Set Membership: |
|