Assay Details
| Gene Symbol: ZNF500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021086689.1 | XP_020942348.1 | 398 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021086689.1 transcripts.
Set Membership: |
|

ZNF500
Genomic Map