Assay Details
| Gene Symbol: PHYHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928841.5 | XP_001928876.1 | 765 | 64 | ||
| XM_013993922.2 | XP_013849376.1 | 1426 | 64 | |||
| GenBank mRNA | AK232373.1 | - | 8 - 9 | 665 | 64 | |
| AK236678.1 | - | 8 - 9 | 660 | 64 | ||
| AK346009.1 | - | 8 - 9 | 658 | 64 | ||
More Information
Set Membership: |
|

PHYHD1
Genomic Map