Assay Details
| Gene Symbol: FLAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001929375.6 | XP_001929410.3 | 1379 | 64 | ||
| XM_003355138.4 | XP_003355186.1 | 1287 | 64 | |||
| XM_021089754.1 | XP_020945413.1 | 2889 | 64 | |||
| GenBank mRNA | AK232502.1 | - | 4 - 5 | 1651 | 64 | |
| AK236650.1 | - | 5 - 6 | 1233 | 64 | ||
| AK346465.1 | - | 5 - 6 | 868 | 64 | ||
More Information
Set Membership: |
|

FLAD1
Genomic Map