Assay Details
| Gene Symbol: LYPLAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021064301.1 | XP_020919960.1 | 152 | 65 | ||
| XM_021064302.1 | XP_020919961.1 | 150 | 65 | |||
| XM_021064303.1 | XP_020919962.1 | 151 | 65 | |||
| XM_021064304.1 | XP_020919963.1 | 151 | 65 | |||
| XM_021064305.1 | XP_020919964.1 | 151 | 65 | |||
| GenBank mRNA | AK232585.1 | - | 1 - 2 | 110 | 65 | |
More Information
Set Membership: |
|

LYPLAL1
Genomic Map