Assay Details
| Gene Symbol: SLC25A30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021065606.1 | XP_020921265.1 | 510 | 79 | ||
| XM_021065607.1 | XP_020921266.1 | 589 | 79 | |||
| GenBank mRNA | AK232610.1 | - | 2 - 3 | 170 | 79 | |
More Information
Set Membership: |
|

SLC25A30
Genomic Map