Assay Details
| Gene Symbol: LIMS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003133402.5 | XP_003133450.3 | 2107 | 72 | ||
| XM_013984126.2 | XP_013839580.1 | 932 | 72 | |||
| XM_021076155.1 | XP_020931814.1 | 1110 | 72 | |||
| XM_021076156.1 | XP_020931815.1 | 1141 | 72 | |||
| XM_021076157.1 | XP_020931816.1 | 869 | 72 | |||
| XM_021076158.1 | XP_020931817.1 | 1018 | 72 | |||
| GenBank mRNA | AK232807.1 | - | 7 - 8 | 874 | 72 | |
More Information
Set Membership: |
|

LIMS2
Genomic Map