Assay Details
| Gene Symbol: TLCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131768.4 | XP_003131816.1 | 401 | 104 | ||
| XM_013989985.2 | XP_013845439.1 | 401 | 104 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: TLCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131768.4 | XP_003131816.1 | 401 | 104 | ||
| XM_013989985.2 | XP_013845439.1 | 401 | 104 | |||
Set Membership: |
|