Assay Details
| Gene Symbol: SLC28A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005659640.3 | XP_005659697.2 | 1236 | 64 | ||
| XM_013993042.2 | XP_013848496.1 | 1133 | 64 | |||
| XM_021096284.1 | XP_020951943.1 | 1236 | 64 | |||
More Information
Set Membership: |
|

SLC28A2
Genomic Map