Assay Details
| Gene Symbol: TAFA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005655438.3 | XP_005655495.1 | 456 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005655438.3 transcripts.
Set Membership: |
|

TAFA3
Genomic Map