Assay Details
| Gene Symbol: LYRM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358165.4 | XP_003358213.1 | 329 | 104 | ||
| XM_005669086.3 | XP_005669143.1 | 799 | 104 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: LYRM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358165.4 | XP_003358213.1 | 329 | 104 | ||
| XM_005669086.3 | XP_005669143.1 | 799 | 104 | |||
Set Membership: |
|