Assay Details
| Gene Symbol: GYG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021079760.1 | XP_020935419.1 | 877 | 58 | ||
| XM_021079761.1 | XP_020935420.1 | 879 | 58 | |||
| XM_021079762.1 | XP_020935421.1 | 820 | 58 | |||
| XM_021079763.1 | XP_020935422.1 | 833 | 58 | |||
| XM_021079764.1 | XP_020935423.1 | 963 | 58 | |||
| XM_021079765.1 | XP_020935424.1 | 877 | 58 | |||
| XM_021079766.1 | XP_020935425.1 | 877 | 58 | |||
| XM_021079767.1 | XP_020935426.1 | 820 | 58 | |||
| XM_021079768.1 | XP_020935427.1 | 853 | 58 | |||
| XM_021079770.1 | XP_020935429.1 | 845 | 58 | |||
More Information
Set Membership: |
|

GYG2
Genomic Map