Assay Details
| Gene Symbol: ZNF33B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013983243.2 | XP_013838697.2 | 871 | 130 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013983243.2 transcripts.
Set Membership: |
|

ZNF33B
Genomic Map