Assay Details
| Gene Symbol: FSIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005659760.3 | XP_005659817.2 | 1242 | 104 | ||
| XM_021097949.1 | XP_020953608.1 | 1373 | 104 | |||
| XM_021097959.1 | XP_020953618.1 | 1375 | 104 | |||
More Information
Set Membership: |
|

FSIP1
Genomic Map