Assay Details
| Gene Symbol: DHRS7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021068046.1 | XP_020923705.1 | 612 | 55 | ||
| XM_021068047.1 | XP_020923706.1 | 663 | 55 | |||
| XM_021068048.1 | XP_020923707.1 | 1085 | 55 | |||
| XM_021068049.1 | XP_020923708.1 | 710 | 55 | |||
| XM_021068050.1 | XP_020923709.1 | 534 | 55 | |||
More Information
Set Membership: |
|

DHRS7B
Genomic Map