Assay Details
| Gene Symbol: TRIM56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021086255.1 | XP_020941914.1 | 170 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021086255.1 transcripts.
Set Membership: |
|

TRIM56
Genomic Map